missing translation for 'onlineSavingsMsg'
Learn More

Vitamin D Receptor (VDR) Ab-1 Rat Monoclonal Antibody, Epredia™

Artikelnummer. p-4559185 Alle ansehen Epredia Produkte
Change view
Click to view available options
Menge:
100 μL
200 μL
500 μL
1 mL
Konjugat:
Biotin
Unconjugated
Konzentration:
1mg/mL
200µg/mL
Packungsgröße:
1 Milliliter
100 Mikroliter
200 Mikroliter
500 Mikroliter
Artikelnummer. Menge Konjugat Konzentration unitSize
12653847 100 μL Biotin 200µg/mL 100 Mikroliter
12663847 500 μL Biotin 200µg/mL 500 Mikroliter
12633847 1 mL Biotin 200µg/mL 1 Milliliter
12683847 100 μL Unconjugated 200µg/mL 100 Mikroliter
12693847 500 μL Unconjugated 200µg/mL 500 Mikroliter
12673847 1 mL Unconjugated 200µg/mL 1 Milliliter
12613857 200 μL Unconjugated 1mg/mL 200 Mikroliter
7 options
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 12653847

Brand: Epredia RT200B0

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

Provide accurate, reproducible results with the Epredia™ Vitamin D Receptor (VDR) Ab-1, Rat Monoclonal Antibody.

VDR is a member of super family of steroid hormone receptors. It is known to play role in cellular growth and differentiation, calcium and phosphorus metabolism and bone remodeling. Human breast carcinomas, leukemias, and prostatic carcinoma cells express VDR, which can modify their growth and differentiation. In avians, VDR appears as a doublet of 58kDa and 60kDa.

Host Species: Rat

Clone: 9A7γ.E10.E4

Isotype: IgG2b

Species Reactivity: Human, Pig, Mouse, Rat, Hamster, Frog, Chicken and Fish. Others not known.

Epitope: aa 89-105 (near the DNA-binding domain)

Immunogen: Vitamin D receptor protein from chicken intestine

Molecular Weight: 53kDa

Positive Control: LNCAP cells, HeLa cells

Cellular Localization: Nuclear Ab-1 reacts with both the occupied and unoccupied forms of VDR and shows no cross reaction with estrogen and glucocorticoid receptors. It is excellent for biochemical and histochemical applications.

Recommended for:

  • Western Blotting
  • Gel Supershift Assay

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Vitamin D Receptor (VDR) Ab-1
Anwendungen Western Blot
Klassifikation Monoclonal
Klon 9A7gamma.E10.E4
Konzentration 200µg/mL
Konjugat Biotin
Wirtsspezies Rat
Immunogen Vitamin D receptor protein from chicken intestine
Menge 100 μL
Regulatorischer Status RUO
Forschungsgebiet Nuclear Receptors
Primär oder sekundär Primary
Zielspezies Chicken, Fish, Frog, Hamster, Human, Mouse, Pig
Form Concentrated
Isotype IgG2b
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.