missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin beta 3 Antibody (CL3363) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-43800
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Laminin beta 3 Monoclonal antibody specifically detects Laminin beta 3 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry
Spezifikation
| Laminin beta 3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| BM600-125KDA, Epiligrin subunit bata, FLJ99565, Kalinin B1 chain, Kalinin subunit beta, kalinin-140kDa, LAM5, Laminin 5, Laminin B1k chain, laminin S B3 chain, laminin subunit beta-3, laminin, beta 3, laminin, beta 3 (nicein (125kD), kalinin (140kD), BM600 (125kD)), Laminin-5 subunit beta, LAMNB1, Nicein subunit beta, nicein-125kDa | |
| This antibody was generated using a recombinant protein sequence of Q13751, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| CL3363 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3914 | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur