Learn More
Thermo Scientific™ LanthaScreen™ Tb-p-c-jun (pSer73) Antibody
Beschreibung
- A part of the activating protein-1 (AP-1) transcription factor, c-Jun is phosphorylated at Ser73 by members of the JNK family
- For use in combination with the GFP-c-Jun physiological protein substrate (PV4446) to evaluate inhibitors in medium to high throughput screening applications
Spezifikation
Spezifikation
| Konjugat | Tb (Terbium)/Lanthanide Chelates |
| Menge | 1 mg |
| Inhalt und Lagerung | Terbium-labeled p-c-Jun (pSer73) antibody in HEPES buffered saline. Store at -20°C. |
| Anregung/Emission | 340/490, 520 |
| Reaktivität | Human |
| Versandbedingung | Dry Ice |
| Anzeigewert | End Point |
| Reaktivität Abk. | Hu |
| Zielgen | Ser/Thr Kinases |
| Produktlinie | LanthaScreen |