Learn More
Thermo Scientific™ LanthaScreen™ Tb-pMAP2K1 (pSer217/221) Antibody
For use in combination with the Fluorescein-MAP2K1 physiological protein substrate (PV4812) to evaluate inhibitiors in medium to high throughput screening applications.
Marke:  Thermo Scientific™ PV4817
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 10480635
  • 4560.00 € / 25 Mikrogramm
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.
Beschreibung
Beschreibung
  • MAP2K1 (mitogen-activated protein kinase kinase 1) is phosphorylated at Ser217/221 by a range of MAPK family members including Raf and B-Raf
  • For use in combination with the Fluorescein-MAP2K1 physiological protein substrate (PV4812) to evaluate inhibitiors in medium to high throughput screening applications
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Tb (Terbium)/Lanthanide Chelates
Terbium-labeled pMAP2K1 (pSer217/221) antibody in HEPES buffered saline. Store at -20°C.
Human
End Point
Ser/Thr Kinases
25 μg
340/490, 520
Dry Ice
Hu
LanthaScreen
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
For Research Use Only. Not for use in diagnostic procedures.