Learn More
Thermo Scientific™ LanthaScreen™ Tb-pMAP2K1 (pSer217/221) Antibody
For use in combination with the Fluorescein-MAP2K1 physiological protein substrate (PV4812) to evaluate inhibitiors in medium to high throughput screening applications.
Marke: Thermo Scientific™ PV4817
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- MAP2K1 (mitogen-activated protein kinase kinase 1) is phosphorylated at Ser217/221 by a range of MAPK family members including Raf and B-Raf
- For use in combination with the Fluorescein-MAP2K1 physiological protein substrate (PV4812) to evaluate inhibitiors in medium to high throughput screening applications
Spezifikation
| Tb (Terbium)/Lanthanide Chelates | |
| Terbium-labeled pMAP2K1 (pSer217/221) antibody in HEPES buffered saline. Store at -20°C. | |
| Human | |
| End Point | |
| Ser/Thr Kinases |
| 25 μg | |
| 340/490, 520 | |
| Dry Ice | |
| Hu | |
| LanthaScreen |