Learn More
Thermo Scientific™ LanthaScreen™ Tb-PY20 Antibody Kit
Beschreibung
A general reagent to monitor tyrosine kinase activity, the pY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate.
For use in combination with Fluorescein-poly-GT (Cat. No. PV3610) or Fluorescein-poly-GAT (PV3611) to evaluate inhibitors in medium to high throughput screening applications.
Spezifikation
Spezifikation
| Antigen | Tyrosine, phospho-20 |
| Konzentration | 0.54mg/mL |
| Konjugat | Tb (Terbium)/FITC |
| Molekulargewicht des Antigens | 150kDa |
| Menge | 1 mg |
| Regulatorischer Status | RUO |
| Forschungsgebiet | Protein Biology |
| Primär oder sekundär | Primary |
| Verpackung | 1 Kit(s) |
| Inhalt und Lagerung | Terbium-labeled anti-pY20 antibody in HEPES buffered saline. Store at -20°C. |
| Mehr anzeigen |