Learn More
Gilac™ Large Volume Container with Pivoting Wheels and Brakes
Click to view available options
Kapazität (metrisch):
100 l
170 l
220 l
310 l
500 l
Enthält:
2 Schwenkrollen mit Bremsen
4 Schwenkrollen, 2 Bremsen
Unit Size:
1 Stück
Description
- The economical solution for moving large volume containers around.
- Stainless steel wheels and fastenings.
- Stainless steel recommended in humid environments (no damage to bearings even in the presence of water).
- Load capacity for information purposes only (depending on how the load is spread and the operating temperature).
- Bottom with gaskets on the axle for perfect sealing.
- Volume in litres is equivalent to the container filled to the brim.
Specifications
Specifications
| Typ | Storage Tote |
| Material | HDPE |
| Farbe | Weiß |
| Form | Rechteckig |
| Kapazität (angloamerikanisch) | 132.1 gal. |
| Kapazität (metrisch) | 500 l |
| Enthält | 2 Schwenkrollen mit Bremsen |