Learn More
Melford LB Broth, Powder
Artikelnummer. 30018413
Change view
Click to view available options
Menge:
100 g
500 g
1 kg
5 kg
Packungsgröße:
1 Kilogramm
100 Gramm
5 Kilogramm
500 Gramm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30018413 100 g 100 Gramm
30018416 5 kg 5 Kilogramm
30018415 500 g 500 Gramm
30018414 1 kg 1 Kilogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 30018413 Lieferant Melford Lieferanten-Nr. L24060100.0
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
For propagation and maintenance of E.
For propagation and maintenance of E. Coli. Low Salt Formula. Molecular Biology Grade.
Dissolve 20 grams in 1 liter of distilled water. Then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Sterilize by autoclaving for 15 minutes.
Ingredients:
Peptone: 10 g/L
Yeast Extract: 5 g/L
Sodium Chloride: 5 g/L
Spezifikation
Menge 100 g