Learn More
LDLR Antibody (C7), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NBP1-78159AF488
Artikelnummer. 18777803
  • 628.00 € / 0.10 Milliliter
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
LDLR Monoclonal specifically detects LDLR in Human, Rat, Porcine, Bovine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
LDLR
Monoclonal
Alexa Fluor 488
P01130
LDLR
Partially purified bovine LDL receptor [Swiss-Prot# P01131].
0.1 mL
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
3949
Store at 4C in the dark.
IgG2b
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
C7
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
Mouse
Protein G purified
RUO
Primary
Human, Rat, Pig, Bovine
Purified
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen