missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Legionella Pneumophila Antibody (2F10), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NB110-2520H
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Legionella Pneumophila Monoclonal specifically detects Legionella Pneumophila in Bacteria samples. It is validated for ELISA, Immunoassay, Sandwich ELISA.
Spezifikation
| Legionella Pneumophila | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with membrane fraction of Legionella pneumophila, strain Philadelphia 1. | |
| 0.2 mL | |
| Primary | |
| Bacteria | |
| Purified |
| ELISA, Immunoassay | |
| 2F10 | |
| ELISA, Immunoassay, Sandwich ELISA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Reacts with LPS membrane of Legionella pneumophila. No cross-reactivity with Legionella pneumophila serogroups 2, 6, 7, 10 and 11, Legionella bozemanii, Legionella lonbeachae, Legionella micdadei, Corynebacterium diphtheria, Pseudomonas fluorescens, Pseudomonas cepacia, Pseudomonas aerugenose, Bordetella pertusis, Leptospira interrogens (Australia, Pomona, Icterogemorrhagia), Toxoplasma gondii, Hemophilus influenza (type B), Hemophilus ducreyi, Brucella abortus, Bacillus anthracis, Klebsiella pneumoniae, Listeria monocytogenes, Mycobacterium tuberculosis, Neisseria meningitides, Pasteurella multocida, Proteus vulgaris, Shigella sonnei, Staphylococcus faecalis, Streptococcus puogenes, Streptococcus pneumoniae, Yersinia pseudotuberculosis, Yersinia pestis EV76, Salmonella typhi, Escherichia coli K88 and Francisella tularensis 15. Cross-reactivity with Legionella pneumophila serogroups 3 and 4 has not been tested. | |
| Store at 4C in the dark. | |
| IgG3 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur