missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Lewis Y Monoclonal antibody specifically detects Lewis Y in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Specifications
Specifications
| Antigen | Lewis Y |
| Anwendungen | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Klassifikation | Monoclonal |
| Klon | A70-A/A9 |
| Konzentration | 0.2 mg/mL |
| Konjugat | Unconjugated |
| Verdünnung | Flow Cytometry 1-2 μg/million cells, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Immunofluorescence 1-2ug/mL |
| Zusammensetzung | 10 mM PBS with 0.05% BSA |
| Gen-Alias | IPCA-2, IPCA6, IPCA-6, IPCA-8, PCANAP2, PCANAP6, PCANAP8, Prostate cancer-associated protein 6, PRST, Solute carrier family 45 member 3 |
| Wirtsspezies | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?