Learn More
Invitrogen™ Lewis Y Biosimilar Recombinant Monoclonal Antibody
GREENER_CHOICE
Artikelnummer. 30283123
Change view
Click to view available options
Menge:
1 mg
100 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
Artikelnummer. Menge unitSize
30283123 100 μg 100 Mikrogramm
30283125 1 mg 1 Milligramm
2 options
Artikelnummer. 30283123
Marke: Invitrogen™ MA558691
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Recombinant Monoclonal Antibody
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term MB 311.
The Lewis Y (Le^y) gene is not actually a single gene but refers to a carbohydrate antigen known as Lewis Y or Le^y antigen, which is part of the Lewis blood group system. This antigen is synthesized by the fucosylation of precursor substances, which involves enzymes encoded by the FUT1 (fucosyltransferase 1) and FUT2 genes. These genes are responsible for the addition of fucose, a hexose deoxy sugar, to the precursor oligosaccharides. The expression of Lewis Y antigen has been extensively studied in the context of cancer biology as it is commonly found on the surface of epithelial and endothelial cells but overexpressed in various types of cancers, including colorectal, gastric, and ovarian carcinomas. The overexpression and altered glycosylation patterns of Le^y antigen in tumors have made it a potential biomarker for cancer diagnosis and a target for immunotherapeutic approaches.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Lewis Y Humanized Biosimilar
Anwendungen ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Klassifikation Recombinant Monoclonal
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Wirtsspezies Human
Reinigungsverfahren Protein A
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Zielspezies Human
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Lyophilized
Isotype IgG1