missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Lgr5/GPR49 Monoclonal antibody specifically detects Lgr5/GPR49 in Human samples. It is validated for Flow Cytometry, Agonist Activity, Immunocytochemistry, CyTOF
Specifications
Specifications
| Antigen | Lgr5/GPR49 |
| Anwendungen | Flow Cytometry, Agonist Activity, Immunocytochemistry, CyTOF |
| Klassifikation | Monoclonal |
| Klon | 707042 |
| Konjugat | DyLight 488 |
| Verdünnung | Flow Cytometry, Agonist Activity, Immunocytochemistry, CyTOF-ready |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | FEX, G protein-coupled receptor 49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38 |
| Wirtsspezies | Mouse |
| Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Lgr5/GPR49, Met1-Ile560, Accession # O75473 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?