missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lgr5/GPR49 Antibody (CL13567) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-24570AF647
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Lgr5/GPR49 Monoclonal antibody specifically detects Lgr5/GPR49 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Spezifikation
| Lgr5/GPR49 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| FEX, G protein-coupled receptor 49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38 | |
| This antibody was developed using a synthetic peptide derived from O75473, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, GPCR, Neuroscience, Signal Transduction, Wnt Signaling Pathway | |
| 8549 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL13567 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur