missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LGR5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533728
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
LGR5 Polyclonal Antibody for ICC/IF, IHC (P)
G protein-coupled receptor GPR49 has been reported to be expressed in brain, skeletal muscle, placenta, and spinal cord. ESTs have been isolated from normal human brain (amygdala), embryo, and placenta libraries and from human germ cell, uterus, and brain libraries. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
Tekniske data
| LGR5 | |
| Polyclonal | |
| Unconjugated | |
| Lgr5 | |
| Fex; follicle expressed hormone; G protein-coupled receptor 49; GPR49; GPR67; G-protein coupled receptor 49; G-protein coupled receptor 67; G-protein coupled receptor HG38; GRP49; HG38; leucine rich repeat containing G protein coupled receptor 5; leucine rich repeat containing G protein-coupled receptor 5; leucine-rich repeat containing G protein-coupled receptor 5; leucine-rich repeat-containing G protein-coupled receptor 5; leucine-rich repeat-containing G-protein coupled receptor 5; Lgr5; orphan G protein-coupled receptor HG38; orphan G-protein coupled receptor FEX | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8549 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 10% glycerol and 0.09% sodium azide | |
| O75473 | |
| Lgr5 | |
| Synthetic 20 amino acid peptide from 1st cytoplasmic domain of human GPR49 / LGR5. Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey (100%); Marmoset, Bovine, Bat (95%); Hamster, Elephant, Panda, Rabbit (90%); Mouse, Rat, Opossum (85%). | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion