Learn More
BOLA Liebig Condensers Vacuum
Click to view available options
Länge (metrisch) Ummantelung:
300 mm
450 mm
Packungsgröße:
1 Stück
Beschreibung
- One-piece cooling tube with ground joint socket and cone size 29 made of PTFE, cooling jacket made of borosilicate glass with hose connectors made of PP and nuts for connection of cooling water.
- The distillate is only exposed to PFA/PTFE. Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.
Tekniske data
Tekniske data
| Zertifizierung/Konformität | FDA |
| Enthält | hose connectors made of PP and nuts for connection |
| Länge (metrisch) Ummantelung | 450 mm |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion