Learn More
Invitrogen™ Lifastuzumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA558882
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Lifastuzumab vedotin is under investigation in clinical trial NCT01363947 (Safety and Pharmacokinetics of Escalating Doses of DNIB0600A in Participants With Non-Small Cell Lung Cancer (NSCLC) and Platinum Resistant Ovarian Cancer).
Spezifikation
| Lifastuzumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| DNIB0600A; MNIB2126A | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |