missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Linavonkibart Recombinant Monoclonal Antibody

Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Linavonkibart, also known as SRK-181, is a fully human IgG4 monoclonal antibody designed to selectively inhibit the latent transforming growth factor-beta isoform 1 (TGFB1). Transforming growth factor-B1 (TGF-B1) is a key member of the TGF-beta family, a group of evolutionarily conserved polypeptide growth factors. The gene encoding TGF-B1 is located on chromosome 19 at the 19q13.2 locus. Structurally, TGF-B1 is synthesized as a precursor, which is then cleaved to produce a mature polypeptide that forms a dimeric protein. Functionally, TGF-B1 plays a crucial role in cellular processes such as proliferation, differentiation, and apoptosis, and is essential for regulating immune responses, maintaining tissue homeostasis, and orchestrating wound healing. Dysregulation of TGF-B1 signaling has been implicated in various pathological conditions including cancer, fibrosis, and autoimmune disorders. The activation of latent TGF-B1 stored in the extracellular matrix is mediated by interaction with integrins and other co-factors that induce conformational changes necessary for its biological activity.
Spezifikation
Spezifikation
| Antigen | Linavonkibart |
| Anwendungen | ELISA, Functional Assay |
| Klassifikation | Recombinant Monoclonal |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 |
| Wirtsspezies | Human |
| Reinigungsverfahren | Protein A |
| Menge | 100 μg |
| Regulatorischer Status | RUO |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?