missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Linavonkibart Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA558788
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Linavonkibart, also known as SRK-181, is a fully human IgG4 monoclonal antibody designed to selectively inhibit the latent transforming growth factor-beta isoform 1 (TGFB1). Transforming growth factor-B1 (TGF-B1) is a key member of the TGF-beta family, a group of evolutionarily conserved polypeptide growth factors. The gene encoding TGF-B1 is located on chromosome 19 at the 19q13.2 locus. Structurally, TGF-B1 is synthesized as a precursor, which is then cleaved to produce a mature polypeptide that forms a dimeric protein. Functionally, TGF-B1 plays a crucial role in cellular processes such as proliferation, differentiation, and apoptosis, and is essential for regulating immune responses, maintaining tissue homeostasis, and orchestrating wound healing. Dysregulation of TGF-B1 signaling has been implicated in various pathological conditions including cancer, fibrosis, and autoimmune disorders. The activation of latent TGF-B1 stored in the extracellular matrix is mediated by interaction with integrins and other co-factors that induce conformational changes necessary for its biological activity.
Spezifikation
| Linavonkibart | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Functional Assay | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG4SP |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur