Learn More
Thermo Scientific™ Lindberg/Blue M™ LGO Box Furnaces
Artikelnummer. 17829393 Alle ansehen Thermo Scientific Produkte
Ansicht ändern
Click to view available options
Kapazität (metrisch):
55,3 l
Temperaturregelung:
Digital Multi Seg/Multi Prog/OTC
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Kapazität (metrisch) Temperaturregelung unitSize
17829393 55,3 l Digital Multi Seg/Multi Prog/OTC 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17829393 Lieferant Thermo Scientific™ Lieferanten-Nr. BF51842PBC1
  • ONLINE EXCLUSIVE PRICE
    13805.00 €
    13200.00 € / 1 Stück
    Sparen 605.00 €
    4% Rabatt
 Eingeschränkter Artikel
Verfügbar ab: 27-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Utilize the latest technical advances in heating elements, insulation and temperature control integrated into a self-contained cabinet with these box furnaces.
Utilize the latest technical advances in heating elements, insulation and temperature control integrated into a self-contained cabinet with Thermo Scientific™ Lindberg/Blue M™ LGO Box Furnaces. Furnaces feature proprietary LGO (light gauge overbend) heating elements and Thermo Scientific™ Moldatherm™ insulation for efficient and economical transfer of heat to chamber, with low exterior temperatures. Variable heat-up rate eliminates thermal shock to materials with inherently quick heat up and cool down rates.
  • Variable heat-up rate eliminates thermal shock to materials with quick heat-up and cool-down rates
  • Choice of side hinge or vertical lift door
  • Air vent (1in. dia., top) and air inlet (0.375in. dia., rear) for inert atmosphere exchange (may experience some leakage at door)
  • Self-tuning, digital instrumentation for precise temperature setpoint and display
  • Platinel II™ thermocouple for long-term stability
  • 0.6 cu. ft. models feature vertical lift door; 2 cu. ft. models feature horizontal side swing door, hot side facing away from operator for protection
Microprocessor Control
  • Microprocessor-based self-tuning PID control (proportional, integral, derivative) provides optimum thermal process, prevents overshoot
  • Control panel designed for easy access and maintenance
  • Main power ON/OFF switch on control panel
  • Adjustable high-limit overtemperature protection
  • Simultaneous LED display of actual temperature vs. setpoint
  • Can be configured to display temperature in either °C or °F
  • Safety door switch interrupts power to heating element when door is opened; protects heating elements and minimizes exposure to end-user
  • Removable shelves for versatility
  • Moldatherm™ hearthplate supports load and prevents damage due to spillage
Digital, Single-Setpoint Controller
  • Single segment, single setpoint, one ramp to setpoint
Digital Single-Program, Multiple-Segment Programmable Controller
  • Single program with multiple segments for ramp (up and down) and dwell (timed hold) temperature control
Digital Multiple-Program, Multiple-Segment Programmable Controller
  • Available on models with
    P designation Multiple programs and segments for ramp (up and down) and dwell (timed hold) temperature control
Overtemperature Control (OTC)†
  • Adjustable digital overtemperature control, available on selected models with
    B suffix designation
  • Protects furnace and load in the event of primary control circuit failure
  • Overrides main controller and shuts off power to furnace if high limit is reached
  • Manual reset required for safety
  • Operates via magnetic contacts through signal from independent thermocouple
Flowmeter Option (FM) (Inert Atmosphere Only)
  • Available on selected models with
    FM designation
  • Gas flowmeter, adjustable, located on front control panel
  • Adjustable flowrate, range 1.0 to 10.0 cu. ft./hr. standard
  • Suitable for inert gas or airflow to chamber
  • Allows fresh air exchange for ashing applications Not suitable for combustible or volatile gases
Ordering Information:
Required power cord and hardwiring not included. Optional RS485 Digital Communications Port allows controller to be connected to a PC for remote monitoring and control of the furnace; up to 30 units can be connected to one PC.
†Specify Option
B for Overtemperature Control (OTC) when ordering.
Includes:
One two-part shelf (0.6 cu. ft. models have one shelf position at center position; 2.0 cu. ft. models have three shelf positions)
TRUSTED_SUSTAINABILITY
Spezifikation
Kapazität (angloamerikanisch) 2.0 cu.ft.
Kapazität (metrisch) 55,3 l
Temperatur (metrisch) 100 °C bis 1200 °C
Typ Multisegment, Multiprogram, OTC
Leistungsaufnahme 5800 w
Temperaturregelung Digital Multi Seg/Multi Prog/OTC
Abmessungen (T x B x H) innen 15 x 15 x 15 in. (38.1 x 38.1 x 38.1 cm)
Abmessungen (L x B x H) außen 28 x 29 x 33 in. (71.1 x 73.7 x 83.8 cm)
Länge (angloamerikanisch) 28 in.
Länge (metrisch) 71.1
Breite (angloamerikanisch) 29 in.
Breite (metrisch) 73.7 cm
Innenbreite (metrisch) 38.1 cm
Höhe (angloamerikanisch) 33 in.
Innenhöhe (angloamerikanisch) 15 in.
Höhe (metrisch) 83.8 cm
Innenhöhe (metrisch) 38.1 cm
Innentiefe (angloamerikanisch) 15 in.
Innentiefe (metrisch) 38.1 cm
Versandgewicht (metrisch) 127 kg
Elektrische Anforderungen 208/240 V 50/60 Hz
Spannung 208/240 V
Frequenz 50/60 Hz