Learn More
Motic™ LM Plan Achromatic Objective
Beschreibung
- Objectives with long working distance for incident light provide optimal image contrast for reflective, non-covered specimen
- Together with a fully corrected tube lens, an intermediate image without colored fringes is created
- 0.8 N.A.
Spezifikation
Spezifikation
| Typ | LM Plan Achromatic Objective |
| Zur Verwendung mit (Geräte) | BA310MET Series |
| Vergrößerungsleistung | 100X |
| Arbeitsabstand | 2 mm |