missing translation for 'onlineSavingsMsg'
Learn More

LRP-4 Antibody, Novus Biologicals™

Artikelnummer. 18497220 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
25ul
Packungsgröße:
0.10 Milliliter
25 Mikroliter
Product Code. Menge unitSize
18497220 25ul 25 Mikroliter
18259195 0.1 mL 0.10 Milliliter
2 options
This item is not returnable. View return policy

Product Code. 18497220

Brand: Novus Biologicals NBP18124025ul

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 1 publication

LRP-4 Polyclonal specifically detects LRP-4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP-4
Anwendungen Immunohistochemistry, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugat Unconjugated
Verdünnung Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Zusammensetzung PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gen-Alias CLSS, KIAA0816, low density lipoprotein receptor-related protein 4, low-density lipoprotein receptor-related protein 4, LRP-4, MEGF7LRP10, Multiple epidermal growth factor-like domains 7
Gensymbole LRP4
Wirtsspezies Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:PGMGNLTYSNPSYRTSTQEVKIEAIPKPAMYNQLCYKKEGGPDHNYTKEKIKIVEGICLLSGDDAEWDDLKQLRSSRGGLLRDHVCMKTDTVSIQASSGSLDDTETEQLLQEEQSECSSVH
Reinigungsverfahren Affinity Purified
Menge 25ul
Regulatorischer Status RUO
Forschungsgebiet Stem Cell Markers
Primär oder sekundär Primary
Gen-ID (Entrez) 4038
Testspezifität Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Zielspezies Human
Inhalt und Lagerung Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.