missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRP8 Recombinant Rabbit Monoclonal Antibody (JE31-73)

Description
LRP8 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Apolipoprotein E receptor 2 (ApoER2) is an NMDA associated protein found in neurons. It participates in important functions from guiding neurons in the embryonic brain to regulating synaptic signaling. It is also involved in debilitating neurodegenerative diseases such as schizophrenia and Alzheimer's disease.
Specifications
Specifications
| Antigen | LRP8 |
| Anwendungen | Immunohistochemistry (Paraffin), Western Blot |
| Klassifikation | Recombinant Monoclonal |
| Klon | JE31-73 |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gen | LRP8 |
| Gen-Zugriffsnummer | Q14114, Q924X6 |
| Gen-Alias | 4932703M08Rik; AA921429; AI848122; ApoE receptor 2; APOER2; Apolipoprotein E receptor 2; HSZ75190; LDL receptor related protein 8; low density lipoprotein receptor relative, 8 ligand binding repeats, brain; low density lipoprotein receptor-related protein 8, apolipoprotein e receptor; low-density lipoprotein receptor-related protein 8; Lr8b; Lrp8; LRP-8; MCI1; Protein LR8B |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?