missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Luciferase Antibody (Luci 21 1-107), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NB600-307PCP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Luciferase Monoclonal specifically detects Luciferase in Firefly samples. It is validated for Flow Cytometry.
Spezifikation
| Luciferase | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Firefly | |
| Purified |
| Flow Cytometry | |
| Luci 21 1-107 | |
| Flow Cytometry | |
| ec 1.13.12.7, Fluc, luciferin 4 monooxygenase | |
| This Luciferase Antibody (Luci 21 1-107) was developed against luciferase protein from Photinus pyralis (North American firefly). [UniProt# P08659]. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Epitope Tags, Fusion Proteins | |
| This Luciferase Antibody (Luci 21 1-107) is specific for Luciferase, recognizing a peptide consisting of the first 258 amino acids. Further epitope mapping has not been done at this time. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only (RUO)
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur