missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lumiliximab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA559424
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Lumiliximab is a chimeric monoclonal antibody which is used as an immunosuppressive pharmaceutical. It is a primatized anti-CD23 macaque/human chimeric antibody that inhibits the production of the IgE antibody, for the potential treatment of allergic conditions. Lumiliximab was developed by IDEC Pharmaceuticals, which was acquired by Biogen. Clinical trials for CLL were terminated in 2010, and for allergic asthma in 2007. Results published from the CLL clinical trial failed to meet primary endpoints. Lumiliximab is a chimeric macaque-human monoclonal antibody to CD23, a protein expressed on virtually all chronic lymphocytic leukemia (CLL) cells. CD23, also known as Fc epsilon RII, or FcepsilonRII, is the low affinity receptor for IgE, an antibody isotype involved in allergy and resistance to parasites and is important in regulation of IgE levels. Unlike many of the antibody receptors, CD23 is a C-type lectin. It is found on mature B cells, activated macrophages, eosinophils, follicular dendritic cells and platelets.
Spezifikation
| Lumiliximab Chimeric | |
| Recombinant Monoclonal | |
| Unconjugated | |
| IDEC-152; P5E8 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur