Learn More
Invitrogen™ Lumretuzumab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559415
 View more versions of this product
Product Code. 30283810
  • 477.00 € / 100 Mikrogramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
This item is not returnable. View return policy
Description
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Lumretuzumab has been used in trials studying the treatment of Neoplasms, Breast Cancer, Squamous Non-Small Cell Lung Cancer, and Non-Squamous Non-Small Cell Lung Cancer.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lumretuzumab Humanized
Recombinant Monoclonal
Unconjugated
RG-7116; RO5479599
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications