Learn More
Novus Biologicals™ Lung Matched Tumor and Normal Tissue Lysate
Artikelnummer. 18195808 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Menge:
2 Fläschchen
Packungsgröße:
2 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18195808 2 Fläschchen 2 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18195808 Lieferant Novus Biologicals™ Lieferanten-Nr. NBP2470962VIAL
Please to purchase this item. Need a web account? Register with us today!
Two matched 1 mg/mL vials (100 μg/vial) of human lung tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.
The Lung Matched Tumor and Normal Tissue Lysate is derived from human.
TRUSTED_SUSTAINABILITY
Spezifikation
Menge 2 Fläschchen
Inhalt und Lagerung Store at -80C. Avoid freeze-thaw cycles.