missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
Ly-6C Monoclonal specifically detects Ly-6C in Mouse samples. It is validated for Flow Cytometry.
Spezifikation
Spezifikation
| Antigen | Ly-6C |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | ER-MP20 |
| Konjugat | PE/ATTO 594 |
| Verdünnung | Flow Cytometry |
| Zusammensetzung | PBS with 0.05% Sodium Azide |
| Gen-Alias | Ly 6c, Ly6c protein, Lymphocyte antigen 6 complex, Lymphocyte antigen 6 complex locus C, Lymphocyte antigen 6C, Lymphocyte antigen Ly 6C |
| Gensymbole | LY6C1 |
| Wirtsspezies | Rat |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?