missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP2-82107SS
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Polyclonal specifically detects Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunocytochemistry.
Spezifikation
| Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| KDM6B | |
| JMJD3 antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human JMJD3. The immunogen is located within amino acids 1370 - 1420 of JMJD3. Amino Acid Squence: KVPGSRTPGHQENNNF | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is specific for Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 and will not recognize other JMJD proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.5 ug/mL, ELISA, Immunocytochemistry/Immunofluorescence 2.5-5 ug/mL | |
| EC 1.14.11, EC 1.14.11.-, JmjC domain-containing protein 3, JMJD3lysine-specific demethylase 6B, jumonji domain containing 3, histone lysine demethylase, Jumonji domain-containing protein 3, KIAA0346jumonji domain containing 3, lysine (K)-specific demethylase 6B, Lysine demethylase 6B | |
| Rabbit | |
| 180 kDa | |
| 0.025 mg | |
| Cancer, Chromatin Research, Transcription Factors and Regulators | |
| 23135 | |
| Rat | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur