Learn More
Thermo Scientific™ Lysine Medium (Dehydrated)
Artikelnummer. 13285709
Change view
Click to view available options
Menge:
500 g
5 kg
Packungsgröße:
500 Gramm
Artikelnummer. Menge unitSize
13285709 5 kg -
10442453 500 g 500 Gramm
2 options
Artikelnummer. 13285709
Marke: Thermo Scientific™ CM0191T
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Isolate and enumerate wild yeasts encountered in brewing while suppressing the pitching yeasts with this specially designed medium.
Isolate and enumerate wild yeasts encountered in brewing while suppressing pitching yeasts with Thermo Scientific™ Oxoid™ Lysine Medium (Dehydrated). This complex, synthetic medium is based on the original formula of Morris and Eddy1 which utilizes the susceptibility of pitching yeasts to lysine to differentiate it from the wild yeast.
Morris and Eddy1 described this complex medium for the isolation and enumeration of wild yeasts in pitching yeast in the brewery industry. Walters and Thiselton2 used a liquid synthetic medium containing lysine as a sole nitrogen source and found that many types of yeast utilize lysine. Later Morris and Eddy1 also formulated solid lysine medium. Most of the Saccharomyces strains used in the brewery industry and other fermentative industries do not use lysine, whereas the wild strains do.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Spezifikation
Form Powder
Produkttyp Dehydrated Culture Media
Menge 5 kg
For Laboratory Use Only