Beschreibung
The MAD2L2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MAD2L2. This antibody recognizes human, mouse, rat antigen. The MAD2L2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spezifikation
Spezifikation
| Antigen | MAD2L2 |
| Anwendungen | Western Blot, Immunohistochemistry, ELISA |
| Klassifikation | Monoclonal |
| Klon | 2A4C2 |
| Konzentration | 1000 μg/mL |
| Konjugat | Unconjugated |
| Wirtsspezies | Mouse |
| Menge | 150 μL |
| Primär oder sekundär | Primary |
| Gen-ID (Entrez) | 10459 |
| Mehr anzeigen |