missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The C11orf85 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| Zelllinie | The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_13232. The protein contains a C-terminal DDK tag. |
| Zur Verwendung mit (Anwendung) | Western Blot |
| Menge | 0,1 mg |
| Lagerungspuffer | RIPA buffer |
| Reinheit | Protein |
| Testspezifität | Homo sapiens C11orf85, mRNA. |
| Inhalt und Lagerung | -80°C. Avoid freeze-thaw cycles. |
| Ziel | MAJIN |
| Protein State | Native |
Nur für Forschungszwecke
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?