Learn More
MAP2 Antibody (CL5420) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NBP2-61417CL1
Artikelnummer. 30287725
  • 1185.00 € / 0.10 Milliliter
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
MAP2 Monoclonal antibody specifically detects MAP2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Single Cell Western,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
MAP2
Monoclonal
CoraFluor 1
DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2
This antibody was developed using a recombinant protein derived from P11137, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers
4133
Store at 4⪚C in the dark. Do not freeze.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Single Cell Western, Immunocytochemistry/Immunofluorescence
CL5420
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen