Learn More
MAP2 Antibody (CL5420), Novus Biologicals™
Artikelnummer. 18071836 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
25 μL
100 μL
Packungsgröße:
100 Mikroliter
25 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18071836 100 μL 100 Mikroliter
18621969 25 μL 25 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18071836 Lieferant Novus Biologicals Lieferanten-Nr. NBP261417
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Mouse Monoclonal Antibody
MAP2 Monoclonal specifically detects MAP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Single Cell Western.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen MAP2
Anwendungen Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Klassifikation Monoclonal
Klon CL5420
Konzentration 1 mg/mL
Konjugat Unconjugated
Verdünnung Western Blot 1 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Single Cell Western 100 ug/ml
Zusammensetzung PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gen-Alias DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2
Gensymbole MAP2
Wirtsspezies Mouse
Immunogen MAP2 Antibody (CL5420) was developed against a human recombinant protein corresponding to amino acids: EAKAPHWTSAPLTEASAHSHPPEIKDQGGAGEGLVRSANGFPYREDEEGAFGEHGSQGTYSNTKENGINGELTSADRETAEEVSARIVQVVTAEAVAVLKGEQEKEAQHKDQTAALPLAAEETANLPPSPPPSPASEQTVT
Molekulargewicht des Antigens 199 kDa
Reinigungsverfahren Protein A purified
Menge 100 μL
Forschungsgebiet Cellular Markers, Cytoskeleton Markers, MAP Kinase Signaling, Neuronal Stem Cells, Neuroscience, Stem Cell Markers
Primär oder sekundär Primary
Gen-ID (Entrez) 4133
Testspezifität Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Zielspezies Human
Inhalt und Lagerung Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1