missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
MAPK1IP1L Polyclonal specifically detects MAPK1IP1L in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spezifikation
Spezifikation
| Antigen | MAPK1IP1L |
| Anwendungen | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Verdünnung | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Zusammensetzung | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gen-Alias | c14_5346, C14orf32, chromosome 14 open reading frame 32, MAPK-interacting and spindle-stabilizing protein, MAPK-interacting and spindle-stabilizing protein-like, MGC23138, MISS, mitogen activated protein kinase 1 interacting protein 1-like, mitogen-activated protein kinase 1 interacting protein 1-like, Mitogen-activated protein kinase 1-interacting protein 1-like |
| Gensymbole | MAPK1IP1L |
| Wirtsspezies | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:APPVPWGTVPPGAWGPPAPYPAPTGSYPTPGLYPTPSNPFQVPSGPSGAPPMPGG |
| Mehr anzeigen |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?