Learn More
Invitrogen™ Marstacimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA559810
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Marstacimab is a fully human monoclonal antibody that binds to and neutralizes TPFI activity and is under development for treatment of hemophilia. Marstacimab is a laboratory-engineered monoclonal antibody designed to treat hemophilia A and B patients, with or without inhibitors. It works by blocking and effectively preventing TFpi from performing the anticoagulant function that it naturally carries out in the human body.
Spezifikation
| Marstacimab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PF-06741086 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |