missing translation for 'onlineSavingsMsg'
Learn More

MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3 Mouse Monoclonal Antibody, Epredia™

Artikelnummer. 12652127 Alle ansehen Epredia Produkte
Change view
Click to view available options
Menge:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Konzentration:
200μg/mL
Packungsgröße:
1 Milliliter
100 Mikroliter
500 Mikroliter
7 Milliliter
Artikelnummer. Menge Form Konzentration unitSize
12652127 500 μL Concentrated 200μg/mL 500 Mikroliter
12642127 100 μL Concentrated 200μg/mL 100 Mikroliter
12632127 1 mL Concentrated 200μg/mL 1 Milliliter
12612137 7 mL Prediluted - 7 Milliliter
4 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Product Code. 12652127

Brand: Epredia MS716P1

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Epredia™ MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.

MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-Ais a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum.

Host Species: Mouse

Clone: M2-7C10 + M2-9E3

Isotype: IgG2b/κ + IgG2b/κ

Species Reactivity: Human and Horse. Others not known.

Epitope: Not determined

Immunogen: Recombinant hMART-1 protein

Molecular Weight: 18kDa

Positive Control: CAC1 melanoma cells and Melanomas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY

Specifications

Antigen MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3
Anwendungen Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Klassifikation Monoclonal
Klon M2-7C10 + M2-9E3
Konzentration 200μg/mL
Konjugat Unconjugated
Wirtsspezies Mouse
Immunogen Recombinant hMART-1 protein
Menge 500 μL
Regulatorischer Status RUO
Forschungsgebiet Cancer and Tumor Biology
Primär oder sekundär Primary
Zielspezies Equine, Human
Form Concentrated
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.