missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MATR3 Recombinant Rabbit Monoclonal Antibody (24GB535)

Artikelnummer. 30275560
Change view
Click to view available options
Menge:
20 μL
100 μL
Packungsgröße:
100 Mikroliter
20 Mikroliter
Artikelnummer. Menge unitSize
30275560 100 μL 100 Mikroliter
30276462 20 μL 20 Mikroliter
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 30275560

Marke: Invitrogen™ MA557464

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Recombinant Monoclonal Antibody

MATR3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

This gene encodes a nuclear matrix protein, which is proposed to stabilize certain messenger RNA species. Mutations of this gene are associated with distal myopathy 2, which often includes vocal cord and pharyngeal weakness. Alternatively spliced transcript variants, including read-through transcripts composed of the upstream small nucleolar RNA host gene 4 (non-protein coding) and matrin 3 gene sequence, have been identified. Pseudogenes of this gene are located on chromosomes 1 and X.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen MATR3
Anwendungen Flow Cytometry, Western Blot, Immunocytochemistry
Klassifikation Recombinant Monoclonal
Klon 24GB535
Konzentration 1.2 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gen MATR3
Gen-Zugriffsnummer P43243, P43244, Q8K310
Gen-Alias 1110061A14Rik; 2810017I02Rik; AI841759; ALS21; AW555618; D030046F20Rik; KIAA0723; MATR3; matrin 3; matrin-3; matrin-3-like protein; mKIAA0723; MPD2; nuclear scaffold protein P130/MAT3; P130/MAT3; VCPDM; vocal cord and pharyngeal weakness with distal myopathy
Gensymbole MATR3
Wirtsspezies Rabbit
Immunogen A synthesized peptide derived from human Matrin 3 (aa 750-800).
Reinigungsverfahren Affinity chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 17184, 29150, 9782
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.