Learn More
BOLA Maxi Propeller Blades
Artikelnummer. 10080772
Change view
Click to view available options
Abmessungen (L x B x H):
140 x 20 x 4 mm
200 x 20 x 6 mm
200 x 26 x 6 mm
Packungsgröße:
1 Stück
Artikelnummer. Abmessungen (L x B x H) unitSize
10080772 140 x 20 x 4 mm 1 Stück
16229621 200 x 20 x 6 mm 1 Stück
10326152 200 x 26 x 6 mm 1 Stück
3 options
Artikelnummer. 10080772
Marke: BOLA C44110
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Maxi Propeller Blades for stainless steel shafts jacketed with PTFE, glass shafts or metal shafts
  • These solid stirrer blades are made of PTFE and have a set of clamp screws made of a PTFE/PEEK compound.
  • The blades can be fixed tightly on BOLA Stirrer Shafts by means of the clamp screws. A spanner wrench is included for easy assembly.
Spezifikation
Abmessungen (L x B x H) 140 x 20 x 4 mm
Beschichtung PTFE