missing translation for 'onlineSavingsMsg'
Learn More

MCC2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

280.00 € - 624.00 €

Spezifikation

Antigen MCC2
Verdünnung Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000
Anwendungen Immunohistochemistry, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugat Unconjugated
Mehr Spezifikationen ansehen

Products 2
Product Code Brand Menge Price Quantity & Availability  
Product Code Brand Menge Price Quantity & Availability  
18460842
View Documents
Novus Biologicals
NBP2-30804-25ul
25 μL
280.00 €
25 Mikroliter
View alternative products
Save Up to 
»
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
18124315
View Documents
Novus Biologicals
NBP2-30804
0.1 mL
624.00 €
0.10 Milliliter
View alternative products
Save Up to 
»
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Description

Description

MCC2 Polyclonal specifically detects MCC2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

MCC2
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
AIE-75 Binding Protein Protein, AIE-75-Binding Protein, AIEBP, MCC-2, Mutated In Colon Cancer Protein 2, USH1C-Binding Protein 1, USHBP1, Usher Syndrome 1C Binding Protein 1, Usher Syndrome Type-1C Protein-Binding Protein 1
USHBP1
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000
Polyclonal
Rabbit
Human
Q8N6Y0
83878
This antibody was developed against a recombinant protein corresponding to amino acids: TLQHTLSSLEAAAAAWRHQPPSHSGPMEFEGTSEGGAGSLGKQEGAGSCQREAARLAERNAWLRLALSSREDELVRTQASLEAIRAEKETLQKEVQELQDSLLRLEPCPHLSHNQAGG
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.