missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
mCherry Polyclonal specifically detects mCherry in Non-species specific samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Live Imaging Microscopy, Knockdown Validated, Fluorescence Imaging.
Spezifikation
Spezifikation
| Anwendungen | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Inhalt und Lagerung | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Isotype | IgG |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?