Learn More
MCL1 Monoclonal Antibody (OTI2D6), TrueMAB™, OriGene
Artikelnummer. 17572941
Encompass_Preferred
Change view
Click to view available options
Menge:
30 μL
100 μL
Packungsgröße:
100 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17572941 100 μL 100 Mikroliter
17592941 30 μL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17572941 Lieferant Origene Technologies Lieferanten-Nr. TA500982
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Mouse Monoclonal Antibody
MCL1 belongs to the Bcl-2 family. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified. The longer gene product (isoform 1) enhances cell survival by inhibiting apoptosis while the alternatively spliced shorter gene product (isoform 2) promotes apoptosis and is death-inducing.
Spezifikation
Antigen MCL1
Anwendungen Immunocytochemistry, Immunofluorescence, Western Blot
Klassifikation Monoclonal
Klon OTI2D6
Konzentration 0.69 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gen MCL1
Gen-Zugriffsnummer Q07820
Gen-Alias BCL2L3, EAT, MCL1-ES, MCL1L, MCL1S, Mcl-1, TM, bcl2-L-3, mcl1/EAT
Gensymbole MCL1
Wirtsspezies Mouse
Immunogen Full length human recombit protein of human MCL1 produced in HEK293T cell.
Reinigungsverfahren Affinity Chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 4170
Zielspezies Human
Inhalt und Lagerung -20° C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotype IgG1