Learn More
MCM-BP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Artikelnummer. p-7208351 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
150 μL
20 μL
Packungsgröße:
150 Mikroliter
20 Mikroliter
Artikelnummer. Menge unitSize
16859114 20 μL 20 Mikroliter
16849114 150 μL 150 Mikroliter
2 options
Artikelnummer. 16859114
Marke: Proteintech 195731AP20UL
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Polyclonal Antibody
MEK1 (Mitogen activated protein kinase kinase 1) catalyzes the concomitant phosphorylation of a threonine and a tyrosine residue in a Thr-Glu-Tyr sequence located in MAP kinases. MEK1 activates ERK1 and ERK2 MAP kinases. Mitogen activated protein kinase kinase 2 (MEK2 or MAPKK2) is a member of a family of tyrosine/threonine protein kinases that activate the ERK1 and 2 and MAPK enzymes by phosphorylating both residues within the threonine/glutamate/tyrosine (TEY) motif in the activation loop. MEK1 and 2 are also activated by dual phosphorylation, which occurs on serine 218 and 222, in the activation loop of the MEK. Threonine 292 of MEK1 is phosphorylated by ERK 2, which serves as a negative feedback loop by suppressing activation of MEK1.
Spezifikation
Antigen MCM-BP
Anwendungen Western Blot
Klassifikation Polyclonal
Konzentration 0.23 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gen MCMBP
Gen-Zugriffsnummer B1H268, Q8R3C0, Q9BTE3
Gen-Alias C10orf119, MCM BP, MCMBP
Gensymbole Mcmbp
Wirtsspezies Rabbit
Immunogen MCM-BP Fusion Protein Ag13750
Reinigungsverfahren Antigen Affinity Chromatography
Menge 20 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 210711, 309009, 79892
Zielspezies Mouse, Human, Rat
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG