missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MCP-1 ELISA Development Kit (TMB), PeproTech®
Click to view available options
Menge:
10 x 96 Tests
Packungsgröße:
1 Stück
Beschreibung
This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.
Spezifikation
Spezifikation
| Zugriffsnummer | P13500 |
| Assay-Bereich | Lot-specific |
| Konjugat | HRP |
| Produkttyp | ELISA |
| Zur Verwendung mit (Geräte) | Colorimetric Microplate Reader |
| Gen-ID (Entrez) | 6347 |
| Gensymbol | Ccl2 |
| Kitinhalt | Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate |
| Menge | 10 x 96 Tests |
| Kennzeichnung | RUO |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu