Learn More
MCPIP1/ZC3H12A Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke:  Novus Biologicals NBP1-76258B
Artikelnummer. 18570294
  • 630.00 € / 0.10 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
MCPIP1/ZC3H12A Polyclonal antibody specifically detects MCPIP1/ZC3H12A in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
MCPIP1/ZC3H12A
Polyclonal
PBS
Rabbit
Peptide affinity purified
RUO
Primary
Human
Purified
Western Blot, ELISA
Biotin
Reg1, Regnase-1, zinc finger CCCH-type containing 12A
Antibody was raised against a 17 amino acid synthetic peptide near the center of human ZC3H12A. The immunogen is located within amino acids 330 - 380 of ZC3H12A. Amino Acid Squence: PPRAPSKDKNGRRPSPS
0.1 mL
Cancer, Cell Biology, Editing and Processing Endonucleases, Immunology, Inflammation
80149
Store at 4°C in the dark.
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen