Learn More
Invitrogen™ MDH1 Recombinant Rabbit Monoclonal Antibody (24GB1295)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557455
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MDH1 Recombinant Monoclonal Antibody for Western Blot, Flow
MDH1 is localized to the cytoplasm and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.
Spezifikation
| MDH1 | |
| Recombinant Monoclonal | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88989, P14152, P40925 | |
| MDH1 | |
| A synthesized peptide derived from human MDH1 (aa 300-334). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1295 | |
| Unconjugated | |
| MDH1 | |
| B230377B03Rik; Cytosolic malate dehydrogenase; D17921; Diiodophenylpyruvate reductase; epididymis secretory protein Li 32; HEL-S-32; malate dehydrogenase (EC 1.1.1.37); malate dehydrogenase 1; malate dehydrogenase 1, NAD (soluble); malate dehydrogenase, cytoplasmic; malate dehydrogenase, cytoplasmic; malate dehydrogenase, peroxisomal; malate dehydrogenase, soluble; Malate dehydrogenase-like enzyme; Mdh; Mdh1; MDHA; Mdhl; MDH-s; MDL1; MGC:1375; Mor2; Mor-2; soluble malate dehydrogenase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17449, 24551, 4190 | |
| -20°C | |
| Liquid |