missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDM2/HDM2 Antibody (SMP14) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 19 publications
Marke: Novus Biologicals NB100-2736SS
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MDM2/HDM2 Monoclonal specifically detects MDM2/HDM2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Spezifikation
| MDM2/HDM2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen 1:200-1:500 | |
| ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| The SMP14 monoclonal antibody also recognises a peptide epitope around Thr-216 of murine MDM2 (when Thr-216 is unphosphorylated) see Zhang & Prives, 2001 (PMID: 11359766) for further details. SMP14 also cross reacts with some cytokeratins (6, 14 & 16). This is only a problem when working with certain epithelial cells and not fibroblasts. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 κ |
| Western Blot | |
| SMP14 | |
| Unconjugated | |
| Q00987 | |
| MDM2 | |
| Synthetic peptide C-SRPSTSSRRRAISE, corresponding to amino acids 154-167 of human MDM2/HDM2. [UniProt# Q00987] | |
| 0.025 mL | |
| p53 Pathway | |
| 4193 | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur