missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MED11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA568219
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Predicted to react with Human and Rat samples.
In mammalian cells, transcription is regulated in part by high molecular weight co-activating complexes that mediate signals between transcriptional activators and RNA polymerase II (Pol II). The mediator complex is one such multiprotein structure that functions as a bridge between regulatory proteins and Pol II, thereby regulating Pol II-dependent transcription. Med11 (mediator complex subunit 11) is a 117 amino acid nuclear protein and component of the mediator complex. The gene encoding Med11 maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Malfunction or loss of p53 expression is associated with malignant cell growth and Li-Fraumeni syndrome. Like p53, BRCA1 is directly involved in DNA repair, and is associated with predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.
Spezifikation
| MED11 | |
| Polyclonal | |
| Unconjugated | |
| MED11 | |
| 1110030J09Rik; AI465144; AW545069; HSPC296; MED11; Mediator complex subunit 11; mediator of RNA polymerase II transcription subunit 11; mediator of RNA polymerase II transcription, subunit 11 homolog; RGD1563202 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 287456, 400569, 66172 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9D8C6, Q9P086 | |
| MED11 | |
| Synthesized peptide derived from human MED11. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?