Learn More
Invitrogen™ MED13L Recombinant Rabbit Monoclonal Antibody (HL2911)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA556786
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MED13L Recombinant Monoclonal Antibody for Western Blot
The protein encoded by this gene is a subunit of the Mediator complex, a large complex of proteins that functions as a transcriptional coactivator for most RNA polymerase II-transcribed genes. The encoded protein is involved in early development of the heart and brain. Defects in this gene are a cause of transposition of the great arteries, dextro-looped (DTGA).[provided by RefSeq, Jul 2010].
Spezifikation
| MED13L | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q71F56 | |
| MED13L | |
| Synthetic peptide of human MED13L. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| HL2911 | |
| Unconjugated | |
| MED13L | |
| 2210413I17Rik; 6330591G05Rik; 9030618F05Rik; Kiaa1025; Med13l; mediator complex subunit 13 like; mediator complex subunit 13-like; Mediator of RNA polymerase II transcription subunit 13-like; mKIAA1025; MRFACD; PROSIT240; Thrap2; thyroid hormone receptor associated protein 2; thyroid hormone receptor-associated protein 2; Thyroid hormone receptor-associated protein complex 240 kDa component-like; TRAP240L | |
| Rabbit | |
| Protein A | |
| RUO | |
| 23389 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |