missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
MED14 Monoclonal Antibody for Western Blot
The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors. This protein contains a bipartite nuclear localization signal. This gene is known to escape chromosome X-inactivation.
Spezifikation
Spezifikation
| Antigen | MED14 |
| Anwendungen | Western Blot |
| Klassifikation | Monoclonal |
| Klon | 483CT11.1.1 |
| Konzentration | Conc. Not Determined |
| Konjugat | Unconjugated |
| Zusammensetzung | Ascites with 0.09% sodium azide; pH 7.4 |
| Gen | Med14 |
| Gen-Zugriffsnummer | A2ABV5, O60244 |
| Gen-Alias | 9930001L01Rik; Activator-recruited cofactor 150 kDa component; ARC150; AU041628; Cofactor required for Sp1 transcriptional activation subunit 2; cofactor required for Sp1 transcriptional activation subunit 2 (150 kDa); cofactor required for Sp1 transcriptional activation, subunit 2; cofactor required for Sp1 transcriptional activation, subunit 2 (150kD); CRSP complex subunit 2; CRSP150; CRSP2; CSRP; CXorf4; DRIP150; ENSMUSG00000073278; EXLM1; Gm641; hRGR1; human homolog of yeast RGR1; MED14; Mediator complex subunit 14; mediator of RNA polymerase II transcription subunit 14; MGC104513; ORF1; RGD1560170; RGR1; RGR1 homolog; thyroid hormone receptor-associated protein complex 170 kDa component; thyroid hormone receptor-associated protein complex component TRAP170; Transcriptional coactivator CRSP150; transcriptional co-activator CRSP150; TRAP170; vitamin D receptor-interacting protein complex component DRIP150; vitamin D3 receptor-interacting protein complex 150 kDa component |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?
